Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGCCCTGGGTAGCAAGAATACT[A/C]TGCTATCTTTACCCTGTTTCAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607743 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRS2 - fibroblast growth factor receptor substrate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042555.2 | Intron | NP_001036020.1 | ||||
NM_001278351.1 | Intron | NP_001265280.1 | ||||
NM_001278353.1 | Intron | NP_001265282.1 | ||||
NM_001278354.1 | Intron | NP_001265283.1 | ||||
NM_001278355.1 | Intron | NP_001265284.1 | ||||
NM_001278356.1 | Intron | NP_001265285.1 | ||||
NM_001278357.1 | Intron | NP_001265286.1 | ||||
NM_006654.4 | Intron | NP_006645.3 | ||||
XM_017018717.1 | Intron | XP_016874206.1 | ||||
XM_017018718.1 | Intron | XP_016874207.1 | ||||
XM_017018719.1 | Intron | XP_016874208.1 |