Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZZZ3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZZZ3 - zinc finger ZZ-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308237.1 | Intron | NP_001295166.1 | ||||
NM_015534.5 | Intron | NP_056349.1 | ||||
XM_005270725.3 | Intron | XP_005270782.1 | ||||
XM_005270726.3 | Intron | XP_005270783.1 | ||||
XM_005270727.3 | Intron | XP_005270784.1 | ||||
XM_005270729.4 | Intron | XP_005270786.1 | ||||
XM_017000970.1 | Intron | XP_016856459.1 | ||||
XM_017000971.1 | Intron | XP_016856460.1 | ||||
XM_017000972.1 | Intron | XP_016856461.1 |