Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTAACGCTTTCTGAAAAAGTG[C/T]CGCCAAATCATCCCAGTCGGAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300616 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LUZP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LUZP4 - leucine zipper protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318840.1 | 83 | UTR 5 | NP_001305769.1 | |||
NM_016383.4 | 83 | Missense Mutation | CCG,TCG | P,S 14 | NP_057467.1 |