Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608793 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPECC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPECC1 - sperm antigen with calponin homology and coiled-coil domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033553.2 | Intron | NP_001028725.1 | ||||
NM_001033554.2 | Intron | NP_001028726.1 | ||||
NM_001033555.2 | Intron | NP_001028727.1 | ||||
NM_001243438.1 | Intron | NP_001230367.1 | ||||
NM_001243439.1 | Intron | NP_001230368.1 | ||||
NM_152904.4 | Intron | NP_690868.3 | ||||
XM_005256860.2 | Intron | XP_005256917.1 | ||||
XM_011524075.1 | Intron | XP_011522377.1 | ||||
XM_017025317.1 | Intron | XP_016880806.1 | ||||
XM_017025318.1 | Intron | XP_016880807.1 | ||||
XM_017025319.1 | Intron | XP_016880808.1 | ||||
XM_017025320.1 | Intron | XP_016880809.1 | ||||
XM_017025321.1 | Intron | XP_016880810.1 | ||||
XM_017025322.1 | Intron | XP_016880811.1 | ||||
XM_017025323.1 | Intron | XP_016880812.1 | ||||
XM_017025324.1 | Intron | XP_016880813.1 | ||||
XM_017025325.1 | Intron | XP_016880814.1 |