Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610924 | ||||||||||||||||||||
Literature Links: |
RBCK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBCK1 - RANBP2-type and C3HC4-type zinc finger containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323956.1 | Intron | NP_001310885.1 | ||||
NM_001323958.1 | Intron | NP_001310887.1 | ||||
NM_001323960.1 | Intron | NP_001310889.1 | ||||
NM_006462.5 | Intron | NP_006453.1 | ||||
NM_031229.3 | Intron | NP_112506.2 | ||||
XM_005260645.1 | Intron | XP_005260702.1 | ||||
XM_011529137.1 | Intron | XP_011527439.1 | ||||
XM_011529138.1 | Intron | XP_011527440.1 | ||||
XM_011529139.2 | Intron | XP_011527441.1 | ||||
XM_011529140.1 | Intron | XP_011527442.1 | ||||
XM_017027594.1 | Intron | XP_016883083.1 | ||||
XM_017027595.1 | Intron | XP_016883084.1 |