Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600039 | ||||||||||||||||||||
Literature Links: |
BCL2L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCL2L1 - BCL2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191.3 | Intron | NP_001182.1 | ||||
NM_001317919.1 | Intron | NP_001304848.1 | ||||
NM_001317920.1 | Intron | NP_001304849.1 | ||||
NM_001317921.1 | Intron | NP_001304850.1 | ||||
NM_001322239.1 | Intron | NP_001309168.1 | ||||
NM_001322240.1 | Intron | NP_001309169.1 | ||||
NM_001322242.1 | Intron | NP_001309171.1 | ||||
NM_138578.2 | Intron | NP_612815.1 | ||||
XM_011528964.2 | Intron | XP_011527266.1 | ||||
XM_017027993.1 | Intron | XP_016883482.1 |