Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605459 MIM: 605460 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCG5 - ATP binding cassette subfamily G member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022436.2 | Intron | NP_071881.1 | ||||
XM_005264480.3 | Intron | XP_005264537.1 | ||||
XM_006712073.3 | Intron | XP_006712136.1 | ||||
XM_006712074.3 | Intron | XP_006712137.1 | ||||
XM_011533024.2 | Intron | XP_011531326.1 | ||||
XM_011533025.2 | Intron | XP_011531327.1 | ||||
XM_011533026.2 | Intron | XP_011531328.1 | ||||
XM_011533027.2 | Intron | XP_011531329.1 | ||||
XM_011533028.2 | Intron | XP_011531330.1 |
ABCG8 - ATP binding cassette subfamily G member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022437.2 | Intron | NP_071882.1 | ||||
XM_005264483.3 | Intron | XP_005264540.1 | ||||
XM_011533029.2 | Intron | XP_011531331.1 | ||||
XM_011533030.2 | Intron | XP_011531332.1 |
DYNC2LI1 - dynein cytoplasmic 2 light intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |