Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604331 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INTS6 - integrator complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039937.1 | 5534 | UTR 3 | NP_001035026.1 | |||
NM_001039938.1 | 5534 | Intron | NP_001035027.1 | |||
NM_001306091.1 | 5534 | UTR 3 | NP_001293020.1 | |||
NM_012141.2 | 5534 | UTR 3 | NP_036273.1 | |||
XM_011535040.2 | 5534 | Intron | XP_011533342.1 | |||
XM_011535041.1 | 5534 | UTR 3 | XP_011533343.1 | |||
XM_017020540.1 | 5534 | UTR 3 | XP_016876029.1 |
SERPINE3 - serpin family E member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101320.1 | 5534 | Intron | NP_001094790.1 | |||
XM_017020707.1 | 5534 | Intron | XP_016876196.1 |
Set Membership: |
HapMap |