Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608148 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SATB2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SATB2 - SATB homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172509.1 | 2966 | UTR 3 | NP_001165980.1 | |||
NM_001172517.1 | 2966 | UTR 3 | NP_001165988.1 | |||
NM_015265.3 | 2966 | UTR 3 | NP_056080.1 | |||
XM_005246396.2 | 2966 | UTR 3 | XP_005246453.1 | |||
XM_006712372.2 | 2966 | UTR 3 | XP_006712435.1 | |||
XM_011510840.2 | 2966 | UTR 3 | XP_011509142.1 | |||
XM_017003656.1 | 2966 | UTR 3 | XP_016859145.1 |
Set Membership: |
HapMap |