Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608101 MIM: 600119 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HILS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HILS1 - histone linker H1 domain, spermatid-specific 1 (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SGCA - sarcoglycan alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000023.3 | Intron | NP_000014.1 | ||||
NM_001135697.2 | Intron | NP_001129169.1 | ||||
XM_011525120.2 | Intron | XP_011523422.2 | ||||
XM_011525121.2 | Intron | XP_011523423.2 | ||||
XM_011525122.2 | Intron | XP_011523424.2 | ||||
XM_011525123.2 | Intron | XP_011523425.2 | ||||
XM_011525124.2 | Intron | XP_011523426.1 | ||||
XM_017024950.1 | Intron | XP_016880439.1 |
Set Membership: |
HapMap |