Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609132 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf234 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf234 - chromosome 1 open reading frame 234 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KDM1A - lysine demethylase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009999.2 | Intron | NP_001009999.1 | ||||
NM_015013.3 | Intron | NP_055828.2 | ||||
XM_005245786.2 | Intron | XP_005245843.1 | ||||
XM_006710472.3 | Intron | XP_006710535.1 | ||||
XM_006710473.2 | Intron | XP_006710536.1 | ||||
XM_006710474.3 | Intron | XP_006710537.1 | ||||
XM_017000716.1 | Intron | XP_016856205.1 | ||||
XM_017000717.1 | Intron | XP_016856206.1 | ||||
XM_017000718.1 | Intron | XP_016856207.1 | ||||
XM_017000719.1 | Intron | XP_016856208.1 | ||||
XM_017000720.1 | Intron | XP_016856209.1 |
Set Membership: |
HapMap |