Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610111 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANO4 - anoctamin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286615.1 | Intron | NP_001273544.1 | ||||
NM_001286616.1 | Intron | NP_001273545.1 | ||||
NM_178826.3 | Intron | NP_849148.2 | ||||
XM_011537911.2 | Intron | XP_011536213.1 | ||||
XM_011537912.2 | Intron | XP_011536214.1 | ||||
XM_011537913.2 | Intron | XP_011536215.1 | ||||
XM_011537914.2 | Intron | XP_011536216.1 | ||||
XM_011537915.2 | Intron | XP_011536217.1 | ||||
XM_011537916.2 | Intron | XP_011536218.1 | ||||
XM_011537918.2 | Intron | XP_011536220.1 | ||||
XM_017018808.1 | Intron | XP_016874297.1 | ||||
XM_017018809.1 | Intron | XP_016874298.1 | ||||
XM_017018810.1 | Intron | XP_016874299.1 | ||||
XM_017018811.1 | Intron | XP_016874300.1 | ||||
XM_017018812.1 | Intron | XP_016874301.1 | ||||
XM_017018813.1 | Intron | XP_016874302.1 | ||||
XM_017018814.1 | Intron | XP_016874303.1 | ||||
XM_017018815.1 | Intron | XP_016874304.1 | ||||
XM_017018816.1 | Intron | XP_016874305.1 | ||||
XM_017018817.1 | Intron | XP_016874306.1 |