Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATATTTTAGAGCTGGTATATTT[A/T]ATTTCTACCTTAACATCAATGATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602486 MIM: 602487 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POP1 - POP1 homolog, ribonuclease P/MRP subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145860.1 | Intron | NP_001139332.1 | ||||
NM_001145861.1 | Intron | NP_001139333.1 | ||||
NM_015029.2 | Intron | NP_055844.2 | ||||
XM_011516801.2 | Intron | XP_011515103.1 |
RIDA - reactive intermediate imine deaminase A homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |