Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191730 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UPP1 - uridine phosphorylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287426.1 | Intron | NP_001274355.1 | ||||
NM_001287428.1 | Intron | NP_001274357.1 | ||||
NM_001287429.1 | Intron | NP_001274358.1 | ||||
NM_001287430.1 | Intron | NP_001274359.1 | ||||
NM_003364.3 | Intron | NP_003355.1 | ||||
XM_005249838.4 | Intron | XP_005249895.1 | ||||
XM_011515512.2 | Intron | XP_011513814.1 | ||||
XM_011515513.2 | Intron | XP_011513815.1 | ||||
XM_011515514.2 | Intron | XP_011513816.1 | ||||
XM_011515515.2 | Intron | XP_011513817.1 |
Set Membership: |
HapMap |