Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCAAATCCACTTGTAAGTGCTG[C/T]TAATTGAAGTGTAGATTCCAGTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF429 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF429 - zinc finger protein 429 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001415.2 | Intron | NP_001001415.2 | ||||
XM_017026748.1 | Intron | XP_016882237.1 | ||||
XM_017026749.1 | Intron | XP_016882238.1 | ||||
XM_017026750.1 | Intron | XP_016882239.1 | ||||
XM_017026751.1 | Intron | XP_016882240.1 | ||||
XM_017026752.1 | Intron | XP_016882241.1 | ||||
XM_017026753.1 | Intron | XP_016882242.1 | ||||
XM_017026754.1 | Intron | XP_016882243.1 | ||||
XM_017026755.1 | Intron | XP_016882244.1 | ||||
XM_017026756.1 | Intron | XP_016882245.1 | ||||
XM_017026757.1 | Intron | XP_016882246.1 |