Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604693 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKAP7 - A-kinase anchoring protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004842.3 | Intron | NP_004833.1 | ||||
NM_016377.3 | Intron | NP_057461.2 | ||||
NM_138633.2 | Intron | NP_619539.1 | ||||
XM_005267228.1 | Intron | XP_005267285.1 | ||||
XM_005267229.4 | Intron | XP_005267286.1 | ||||
XM_006715614.2 | Intron | XP_006715677.1 | ||||
XM_011536262.1 | Intron | XP_011534564.1 | ||||
XM_017011506.1 | Intron | XP_016866995.1 | ||||
XM_017011507.1 | Intron | XP_016866996.1 | ||||
XM_017011508.1 | Intron | XP_016866997.1 | ||||
XM_017011509.1 | Intron | XP_016866998.1 | ||||
XM_017011510.1 | Intron | XP_016866999.1 | ||||
XM_017011511.1 | Intron | XP_016867000.1 |
Set Membership: |
HapMap |