Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603791 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC23A2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC23A2 - solute carrier family 23 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005116.5 | 4649 | UTR 3 | NP_005107.4 | |||
NM_203327.1 | 4649 | UTR 3 | NP_976072.1 | |||
XM_011529414.2 | 4649 | UTR 3 | XP_011527716.1 | |||
XM_011529415.2 | 4649 | UTR 3 | XP_011527717.1 | |||
XM_011529416.2 | 4649 | UTR 3 | XP_011527718.1 | |||
XM_011529417.2 | 4649 | UTR 3 | XP_011527719.1 | |||
XM_017028171.1 | 4649 | UTR 3 | XP_016883660.1 | |||
XM_017028172.1 | 4649 | UTR 3 | XP_016883661.1 | |||
XM_017028173.1 | 4649 | UTR 3 | XP_016883662.1 | |||
XM_017028174.1 | 4649 | UTR 3 | XP_016883663.1 | |||
XM_017028175.1 | 4649 | UTR 3 | XP_016883664.1 | |||
XM_017028176.1 | 4649 | UTR 3 | XP_016883665.1 | |||
XM_017028177.1 | 4649 | UTR 3 | XP_016883666.1 | |||
XM_017028178.1 | 4649 | UTR 3 | XP_016883667.1 | |||
XM_017028179.1 | 4649 | UTR 3 | XP_016883668.1 | |||
XM_017028180.1 | 4649 | UTR 3 | XP_016883669.1 | |||
XM_017028181.1 | 4649 | Intron | XP_016883670.1 |
Set Membership: |
HapMap |