Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGTACCAGGAGGCCAGAGCCTG[A/T]CACTCTCGCCTCATTCAGTGCTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF718 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF718 - zinc finger protein 718 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039127.5 | 97 | Intron | NP_001034216.2 | |||
NM_001289930.1 | 97 | Intron | NP_001276859.1 | |||
NM_001289931.1 | 97 | Intron | NP_001276860.1 | |||
XM_017007979.1 | 97 | Intron | XP_016863468.1 | |||
XM_017007980.1 | 97 | UTR 5 | XP_016863469.1 | |||
XM_017007981.1 | 97 | Intron | XP_016863470.1 |
Set Membership: |
HapMap |