Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTCACACCTTTTTCAAAGACA[T/G]ACAGCGTGACCTGAGAGGTCTTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 159440 MIM: 602413 | ||||||||||||||||||||
Literature Links: |
MPZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPZ - myelin protein zero | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000530.7 | 498 | Missense Mutation | TAT,TCT | Y,S 145 | NP_000521.2 | |
NM_001315491.1 | 498 | Missense Mutation | TAT,TCT | Y,S 145 | NP_001302420.1 | |
XM_017001321.1 | 498 | Missense Mutation | TAT,TCT | Y,S 155 | XP_016856810.1 |
SDHC - succinate dehydrogenase complex subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |