Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACAACAGGAAAAGCCCAGAAC[A/G]CTTCCTGGAGATAGCGAGGGGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605546 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GP6 - glycoprotein VI platelet | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDH13 - retinol dehydrogenase 13 (all-trans/9-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145971.1 | 1698 | Intron | NP_001139443.1 | |||
NM_138412.3 | 1698 | Intron | NP_612421.1 | |||
XM_005258473.1 | 1698 | Intron | XP_005258530.1 | |||
XM_011526407.1 | 1698 | Intron | XP_011524709.1 | |||
XM_011526408.2 | 1698 | UTR 3 | XP_011524710.1 |
Set Membership: |
HapMap |