Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605792 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT4-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEPT4-AS1 - SEPT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX14 - testis expressed 14, intercellular bridge forming factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201457.1 | 4513 | Silent Mutation | AAA,AAG | K,K 1437 | NP_001188386.1 | |
NM_031272.4 | 4513 | Silent Mutation | AAA,AAG | K,K 1391 | NP_112562.3 | |
NM_198393.3 | 4513 | Silent Mutation | AAA,AAG | K,K 1431 | NP_938207.2 | |
XM_011525029.2 | 4513 | Silent Mutation | AAA,AAG | K,K 1472 | XP_011523331.1 | |
XM_017024860.1 | 4513 | Silent Mutation | AAA,AAG | K,K 1250 | XP_016880349.1 | |
XM_017024861.1 | 4513 | Silent Mutation | AAA,AAG | K,K 1039 | XP_016880350.1 |