Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACTGCATTCCAGCCTCTAAAA[A/G]AACAAAACAAAACAAAACAAACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604128 | ||||||||||||||||||||
Literature Links: |
TCEA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCEA3 - transcription elongation factor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003196.2 | 2261 | Intron | NP_003187.1 | |||
XM_006710864.1 | 2261 | Intron | XP_006710927.1 | |||
XM_011542053.2 | 2261 | UTR 3 | XP_011540355.1 | |||
XM_017002200.1 | 2261 | Intron | XP_016857689.1 | |||
XM_017002201.1 | 2261 | Intron | XP_016857690.1 | |||
XM_017002202.1 | 2261 | Intron | XP_016857691.1 | |||
XM_017002203.1 | 2261 | Intron | XP_016857692.1 | |||
XM_017002204.1 | 2261 | Intron | XP_016857693.1 |
ZNF436-AS1 - ZNF436 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |