Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609093 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO15 - F-box protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142958.1 | Intron | NP_001136430.1 | ||||
NM_152676.2 | Intron | NP_689889.1 | ||||
XM_011525856.1 | Intron | XP_011524158.1 | ||||
XM_011525858.2 | Intron | XP_011524160.1 | ||||
XM_011525859.1 | Intron | XP_011524161.1 | ||||
XM_011525860.1 | Intron | XP_011524162.1 | ||||
XM_017025603.1 | Intron | XP_016881092.1 | ||||
XM_017025604.1 | Intron | XP_016881093.1 |