Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601193 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRISP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRISP1 - cysteine rich secretory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131.2 | Intron | NP_001122.2 | ||||
NM_001205220.1 | Intron | NP_001192149.1 | ||||
NM_170609.1 | Intron | NP_733758.1 | ||||
XM_011514315.2 | Intron | XP_011512617.1 | ||||
XM_017010320.1 | Intron | XP_016865809.1 | ||||
XM_017010321.1 | Intron | XP_016865810.1 |
Set Membership: |
HapMap |