Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGAATAGCATATTAGTAAATAG[A/T]ACCCAAAAGAATAATTCACAATGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120324 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL14A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COL14A1 - collagen type XIV alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021110.3 | Intron | NP_066933.1 | ||||
XM_005251059.3 | Intron | XP_005251116.1 | ||||
XM_006716651.3 | Intron | XP_006716714.1 | ||||
XM_017013809.1 | Intron | XP_016869298.1 |
Set Membership: |
HapMap |