Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAAGAACCTGGCTGGTGCAGCC[A/C]CCAAGGGTGATAAGAGACACATGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606184 | ||||||||||||||||||||
Literature Links: |
ADAMTS12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS12 - ADAM metallopeptidase with thrombospondin type 1 motif 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324511.1 | Intron | NP_001311440.1 | ||||
NM_001324512.1 | Intron | NP_001311441.1 | ||||
NM_030955.3 | Intron | NP_112217.2 | ||||
XM_017009905.1 | Intron | XP_016865394.1 | ||||
XM_017009906.1 | Intron | XP_016865395.1 | ||||
XM_017009907.1 | Intron | XP_016865396.1 | ||||
XM_017009908.1 | Intron | XP_016865397.1 | ||||
XM_017009909.1 | Intron | XP_016865398.1 |