Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACTTCAACAAGCTGTCTTAGAG[C/T]GGTGTATCTGGGTTTACCTTATGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602567 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCC2-AS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GCC2-AS1 - GCC2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIMS1 - LIM zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193482.1 | Intron | NP_001180411.1 | ||||
NM_001193483.2 | Intron | NP_001180412.1 | ||||
NM_001193484.1 | Intron | NP_001180413.1 | ||||
NM_001193485.2 | Intron | NP_001180414.1 | ||||
NM_001193488.1 | Intron | NP_001180417.1 | ||||
NM_004987.5 | Intron | NP_004978.2 | ||||
XM_005263946.1 | Intron | XP_005264003.1 | ||||
XM_005263948.3 | Intron | XP_005264005.1 | ||||
XM_005263949.1 | Intron | XP_005264006.1 | ||||
XM_005263950.1 | Intron | XP_005264007.1 | ||||
XM_005263951.4 | Intron | XP_005264008.1 | ||||
XM_017004091.1 | Intron | XP_016859580.1 | ||||
XM_017004092.1 | Intron | XP_016859581.1 | ||||
XM_017004093.1 | Intron | XP_016859582.1 | ||||
XM_017004094.1 | Intron | XP_016859583.1 | ||||
XM_017004095.1 | Intron | XP_016859584.1 | ||||
XM_017004096.1 | Intron | XP_016859585.1 |
Set Membership: |
HapMap |