Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCATTAATTCTGTTTGAGAAAC[A/T]TTTAAGAATCAATAAAGTATGCCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614047 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPGN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPGN - epithelial mitogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270989.1 | Intron | NP_001257918.1 | ||||
NM_001270990.1 | Intron | NP_001257919.1 | ||||
NM_001270991.1 | Intron | NP_001257920.1 | ||||
NM_001270992.1 | Intron | NP_001257921.1 | ||||
NM_001270993.1 | Intron | NP_001257922.1 | ||||
XM_005265666.4 | Intron | XP_005265723.1 | ||||
XM_011531817.2 | Intron | XP_011530119.1 |
LOC105377276 - uncharacterized LOC105377276 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTHFD2L - methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |