Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGTGATTTTTCTGTCTGTATAC[A/G]CAGTGATAATTCATTTAGATTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC13A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC13A1 - solute carrier family 13 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324400.1 | 3246 | UTR 3 | NP_001311329.1 | |||
NM_022444.3 | 3246 | UTR 3 | NP_071889.2 | |||
XM_011516515.2 | 3246 | UTR 3 | XP_011514817.1 | |||
XM_011516516.2 | 3246 | UTR 3 | XP_011514818.1 | |||
XM_011516517.2 | 3246 | UTR 3 | XP_011514819.1 | |||
XM_011516518.2 | 3246 | Intron | XP_011514820.1 | |||
XM_011516519.2 | 3246 | Intron | XP_011514821.1 | |||
XM_017012554.1 | 3246 | UTR 3 | XP_016868043.1 | |||
XM_017012555.1 | 3246 | Intron | XP_016868044.1 |