Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300003 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARSF - arylsulfatase F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201538.1 | Intron | NP_001188467.1 | ||||
NM_001201539.1 | Intron | NP_001188468.1 | ||||
NM_004042.4 | Intron | NP_004033.2 | ||||
XM_011545522.1 | Intron | XP_011543824.1 | ||||
XM_011545523.1 | Intron | XP_011543825.1 | ||||
XM_011545524.1 | Intron | XP_011543826.1 | ||||
XM_017029527.1 | Intron | XP_016885016.1 | ||||
XM_017029528.1 | Intron | XP_016885017.1 |
Set Membership: |
HapMap |