Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607659 MIM: 609237 | |||||||||||||||||||||||
Literature Links: |
EAF2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EAF2 - ELL associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320041.1 | 418 | UTR 5 | NP_001306970.1 | |||
NM_018456.5 | 418 | Silent Mutation | GGC,GGT | G,G 54 | NP_060926.2 | |
XM_005247618.3 | 418 | Silent Mutation | GGC,GGT | G,G 65 | XP_005247675.1 | |
XM_017006861.1 | 418 | UTR 5 | XP_016862350.1 | |||
XM_017006862.1 | 418 | UTR 5 | XP_016862351.1 | |||
XM_017006863.1 | 418 | Silent Mutation | GGC,GGT | G,G 54 | XP_016862352.1 |
IQCB1 - IQ motif containing B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |