Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGGAGAGCTGCCTGCAGGAGGG[A/C]CCCTCCCTCAAACTCTGGTTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STX2 - syntaxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001980.3 | Intron | NP_001971.2 | ||||
NM_194356.2 | Intron | NP_919337.1 | ||||
XM_005253557.1 | Intron | XP_005253614.1 | ||||
XM_017018978.1 | Intron | XP_016874467.1 | ||||
XM_017018979.1 | Intron | XP_016874468.1 | ||||
XM_017018980.1 | Intron | XP_016874469.1 | ||||
XM_017018981.1 | Intron | XP_016874470.1 | ||||
XM_017018982.1 | Intron | XP_016874471.1 | ||||
XM_017018983.1 | Intron | XP_016874472.1 | ||||
XM_017018984.1 | Intron | XP_016874473.1 | ||||
XM_017018985.1 | Intron | XP_016874474.1 | ||||
XM_017018986.1 | Intron | XP_016874475.1 | ||||
XM_017018987.1 | Intron | XP_016874476.1 | ||||
XM_017018988.1 | Intron | XP_016874477.1 | ||||
XM_017018989.1 | Intron | XP_016874478.1 |
Set Membership: |
HapMap |