Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCGCAAATCTCGTGGCATCTCT[C/T]AAGTCAGTAGTAAAGAAAGTTAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102642 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SOAT1 - sterol O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252511.1 | 207 | Intron | NP_001239440.1 | |||
NM_001252512.1 | 207 | Intron | NP_001239441.1 | |||
NM_003101.5 | 207 | Intron | NP_003092.4 | |||
XM_011509911.1 | 207 | UTR 5 | XP_011508213.1 | |||
XM_011509912.1 | 207 | Intron | XP_011508214.1 |
Set Membership: |
HapMap |