Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCGAGTTCTGGGTGGAGCTCAA[C/T]ACCTACAAGGCCAACGGCAGCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606898 MIM: 606900 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNG6 - calcium voltage-gated channel auxiliary subunit gamma 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031897.2 | 1418 | Silent Mutation | AAC,AAT | N,N 66 | NP_114103.2 | |
NM_145814.1 | 1418 | Silent Mutation | AAC,AAT | N,N 66 | NP_665813.1 | |
NM_145815.1 | 1418 | Silent Mutation | AAC,AAT | N,N 66 | NP_665814.1 | |
XM_006723316.3 | 1418 | Silent Mutation | AAC,AAT | N,N 66 | XP_006723379.1 |
CACNG8 - calcium voltage-gated channel auxiliary subunit gamma 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |