Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604048 | |||||||||||||||||||||||
Literature Links: |
SLC22A14 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC22A14 - solute carrier family 22 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320033.1 | Intron | NP_001306962.1 | ||||
NM_004803.4 | Intron | NP_004794.2 | ||||
XM_005265585.4 | Intron | XP_005265642.1 | ||||
XM_006713416.3 | Intron | XP_006713479.2 | ||||
XM_006713417.3 | Intron | XP_006713480.2 | ||||
XM_006713418.3 | Intron | XP_006713481.2 | ||||
XM_011534245.2 | Intron | XP_011532547.1 | ||||
XM_011534247.2 | Intron | XP_011532549.1 |