Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGGTCATCGCGCTGACGGCGGC[G/T]GCCAACTCGCTGCTGATCGCGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601109 MIM: 600613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTR6 - 5-hydroxytryptamine receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000871.2 | 593 | Silent Mutation | GCG,GCT | A,A 42 | NP_000862.1 |
MINOS1-NBL1 - MINOS1-NBL1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NBL1 - neuroblastoma 1, DAN family BMP antagonist | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |