Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCTTGCCCTTTGTCTCTTGAG[G/T]ACACAGCAAGAAGTCTGTGAGCTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602901 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNPO1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TNPO1 - transportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002270.3 | Intron | NP_002261.3 | ||||
NM_153188.2 | Intron | NP_694858.1 | ||||
XM_005248500.2 | Intron | XP_005248557.1 | ||||
XM_005248501.1 | Intron | XP_005248558.1 | ||||
XM_017009458.1 | Intron | XP_016864947.1 |
Set Membership: |
HapMap |