Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCAGTCCTGGCTGAGGAACTCA[C/G]TGGCCCACTGGGCAGAGATAATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600422 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FABP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FABP6 - fatty acid binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040442.1 | Intron | NP_001035532.1 | ||||
NM_001130958.1 | Intron | NP_001124430.1 | ||||
NM_001445.2 | Intron | NP_001436.1 |