Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610620 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADPRHL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADPRHL1 - ADP-ribosylhydrolase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCUN1D2 - defective in cullin neddylation 1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014283.1 | 6083 | UTR 3 | NP_001014305.1 | |||
XM_017020643.1 | 6083 | UTR 3 | XP_016876132.1 | |||
XM_017020644.1 | 6083 | UTR 3 | XP_016876133.1 | |||
XM_017020645.1 | 6083 | UTR 3 | XP_016876134.1 | |||
XM_017020646.1 | 6083 | UTR 3 | XP_016876135.1 | |||
XM_017020647.1 | 6083 | UTR 3 | XP_016876136.1 | |||
XM_017020648.1 | 6083 | UTR 3 | XP_016876137.1 | |||
XM_017020649.1 | 6083 | UTR 3 | XP_016876138.1 |