Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 172100 | ||||||||||||||||||||
Literature Links: |
PGM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGM3 - phosphoglucomutase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199917.1 | 231 | Intron | NP_001186846.1 | |||
NM_001199918.1 | 231 | Intron | NP_001186847.1 | |||
NM_001199919.1 | 231 | Intron | NP_001186848.1 | |||
NM_015599.2 | 231 | Intron | NP_056414.1 | |||
XM_017010935.1 | 231 | Intron | XP_016866424.1 | |||
XM_017010936.1 | 231 | Intron | XP_016866425.1 | |||
XM_017010937.1 | 231 | Intron | XP_016866426.1 | |||
XM_017010938.1 | 231 | Intron | XP_016866427.1 |
RWDD2A - RWD domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322335.1 | 231 | UTR 5 | NP_001309264.1 | |||
NM_001322336.1 | 231 | UTR 5 | NP_001309265.1 | |||
NM_001322337.1 | 231 | UTR 5 | NP_001309266.1 | |||
NM_001322339.1 | 231 | UTR 5 | NP_001309268.1 | |||
NM_033411.4 | 231 | Missense Mutation | ACA,ATA | T,I 60 | NP_219479.2 | |
XM_017010222.1 | 231 | UTR 5 | XP_016865711.1 | |||
XM_017010223.1 | 231 | UTR 5 | XP_016865712.1 |