Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603948 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM8A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM8A1 - family with sequence similarity 8 member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP153 - nucleoporin 153 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278209.1 | Intron | NP_001265138.1 | ||||
NM_001278210.1 | Intron | NP_001265139.1 | ||||
NM_005124.3 | Intron | NP_005115.2 | ||||
XM_005249507.2 | Intron | XP_005249564.1 | ||||
XM_006715290.2 | Intron | XP_006715353.1 | ||||
XM_006715291.3 | Intron | XP_006715354.1 | ||||
XM_011515028.2 | Intron | XP_011513330.1 | ||||
XM_017011594.1 | Intron | XP_016867083.1 |
Set Membership: |
HapMap |