Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604495 MIM: 612804 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCER2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCER2 - coiled-coil glutamate rich protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBIB - NFKB inhibitor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SARS2 - seryl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145901.1 | Intron | NP_001139373.1 | ||||
NM_017827.3 | Intron | NP_060297.1 |
Set Membership: |
HapMap |