Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCTAACAAATAAATAAGAAAA[C/T]AGGATAGACTGCTTAATGATGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182530 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGEF33 - Rho guanine nucleotide exchange factor 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOS1 - SOS Ras/Rac guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005633.3 | 6055 | UTR 3 | NP_005624.2 | |||
XM_005264515.4 | 6055 | UTR 3 | XP_005264572.1 | |||
XM_011533062.2 | 6055 | UTR 3 | XP_011531364.1 | |||
XM_011533064.2 | 6055 | UTR 3 | XP_011531366.1 | |||
XM_011533066.2 | 6055 | UTR 3 | XP_011531368.1 |
Set Membership: |
HapMap |