Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107941 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRB2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARRB2 - arrestin beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257328.1 | 474 | Silent Mutation | ATC,ATT | I,I 82 | NP_001244257.1 | |
NM_001257329.1 | 474 | Silent Mutation | ATC,ATT | I,I 82 | NP_001244258.1 | |
NM_001257330.1 | 474 | Silent Mutation | ATC,ATT | I,I 82 | NP_001244259.1 | |
NM_001257331.1 | 474 | Silent Mutation | ATC,ATT | I,I 67 | NP_001244260.1 | |
NM_004313.3 | 474 | Silent Mutation | ATC,ATT | I,I 82 | NP_004304.1 | |
NM_199004.1 | 474 | Silent Mutation | ATC,ATT | I,I 67 | NP_945355.1 | |
XM_006721520.1 | 474 | UTR 5 | XP_006721583.1 | |||
XM_011523858.2 | 474 | Silent Mutation | ATC,ATT | I,I 113 | XP_011522160.1 | |
XM_017024645.1 | 474 | UTR 5 | XP_016880134.1 |
Set Membership: |
HapMap |