Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGTCTACTTATATCTGTGTTGC[C/G]TCGGGTTTTTCTTTTTAGGCAGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606957 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF257 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF257 - zinc finger protein 257 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316996.1 | Intron | NP_001303925.1 | ||||
NM_001316997.1 | Intron | NP_001303926.1 | ||||
NM_001316998.1 | Intron | NP_001303927.1 | ||||
NM_033468.3 | Intron | NP_258429.2 |
Set Membership: |
HapMap |