Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCAAATTCTAAAAAGGGAATAAA[C/T]GAATTTTCAGGGTATCAAGCCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615787 MIM: 616391 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NADK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NADK2 - NAD kinase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RANBP3L - RAN binding protein 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161429.2 | 2169 | Intron | NP_001154901.1 | |||
NM_001323273.1 | 2169 | Intron | NP_001310202.1 | |||
NM_001323274.1 | 2169 | UTR 3 | NP_001310203.1 | |||
NM_001323275.1 | 2169 | Intron | NP_001310204.1 | |||
NM_001323276.1 | 2169 | UTR 3 | NP_001310205.1 | |||
NM_001323277.1 | 2169 | UTR 3 | NP_001310206.1 | |||
NM_001323278.1 | 2169 | Intron | NP_001310207.1 | |||
NM_001323279.1 | 2169 | Intron | NP_001310208.1 | |||
NM_001323280.1 | 2169 | UTR 3 | NP_001310209.1 | |||
NM_145000.4 | 2169 | Intron | NP_659437.3 | |||
XM_006714452.3 | 2169 | Intron | XP_006714515.1 | |||
XM_006714453.3 | 2169 | Intron | XP_006714516.1 | |||
XM_006714455.3 | 2169 | Intron | XP_006714518.1 |
Set Membership: |
HapMap |