Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608304 | ||||||||||||||||||||
Literature Links: |
CAGE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAGE1 - cancer antigen 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIOK1 - RIO kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031480.2 | Intron | NP_113668.2 | ||||
NM_153005.1 | Intron | NP_694550.1 | ||||
XM_005249446.3 | Intron | XP_005249503.1 | ||||
XM_011514933.2 | Intron | XP_011513235.2 |