Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCAAAACCAATCCTTCAGGGC[A/G]AAAATGGATGACGCTATTGAACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610586 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP24 - Rho GTPase activating protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025616.2 | Intron | NP_001020787.2 | ||||
NM_001042669.1 | Intron | NP_001036134.1 | ||||
NM_001287805.1 | Intron | NP_001274734.1 | ||||
NM_031305.2 | Intron | NP_112595.2 | ||||
XM_011532300.2 | Intron | XP_011530602.1 | ||||
XM_017008679.1 | Intron | XP_016864168.1 |