Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTATCTTCCTGGAGCTTAGCCG[A/C]GTCTTCCTGGAGCTGAGAGGTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ILF3 - interleukin enhancer binding factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137673.1 | Intron | NP_001131145.1 | ||||
NM_004516.3 | Intron | NP_004507.2 | ||||
NM_012218.3 | Intron | NP_036350.2 | ||||
NM_017620.2 | Intron | NP_060090.2 | ||||
NM_153464.2 | Intron | NP_703194.1 | ||||
XM_006722742.2 | Intron | XP_006722805.1 | ||||
XM_011527984.2 | Intron | XP_011526286.2 | ||||
XM_011527985.2 | Intron | XP_011526287.1 | ||||
XM_011527987.1 | Intron | XP_011526289.1 | ||||
XM_017026763.1 | Intron | XP_016882252.1 | ||||
XM_017026764.1 | Intron | XP_016882253.1 | ||||
XM_017026765.1 | Intron | XP_016882254.1 |
ILF3-AS1 - ILF3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |