Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601199 | |||||||||||||||||||||||
Literature Links: |
CASR PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CASR - calcium sensing receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000388.3 | 362 | Intron | NP_000379.2 | |||
NM_001178065.1 | 362 | Intron | NP_001171536.1 | |||
XM_005247837.2 | 362 | Intron | XP_005247894.1 | |||
XM_006713789.3 | 362 | UTR 5 | XP_006713852.1 | |||
XM_017007324.1 | 362 | Intron | XP_016862813.1 | |||
XM_017007325.1 | 362 | Intron | XP_016862814.1 |